AMBH2D6F7D83 Ambeed, Inc.

5-(2-Fluorophenyl)-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD11875867
  • Cas Number : 881674-56-2
  • Molecular Weight : 189.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/881674-56-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/881674-56-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart