AMBH2D6F7DFA Ambeed, Inc.

(5-(Pyridin-3-yl)-1H-pyrazol-3-yl)methanol

  • MDL number MFCD20527516
  • Cas Number : 287494-03-5
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/287494-03-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/287494-03-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart