AMBH2D6F7E0C Ambeed, Inc.

(S)-6-Amino-2-((S)-2-((S)-6-amino-2-palmitamidohexanamido)-3-methylbutanamido)hexanoic acid compound with 2,2,2-trifluoroacetic acid (1:2)

  • MDL number MFCD16621111
  • Cas Number : 623172-56-5
  • Molecular Weight : 839.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/623172-56-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/623172-56-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart