AMBH2D6F7E2D Ambeed, Inc.

5-Fluorobenzo[d]isoxazol-3(2H)-one

  • MDL number MFCD11846402
  • Cas Number : 99822-23-8
  • Molecular Weight : 153.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/99822-23-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/99822-23-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart