AMBH2D6F7E41 Ambeed, Inc.

4-Chloro-2-vinylpyridine

  • MDL number MFCD15475064
  • Cas Number : 98420-89-4
  • Molecular Weight : 139.5800

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form liquid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/98420-89-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/98420-89-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart