AMBH2D6F7E48 Ambeed, Inc.

5-Bromo-2,3-dichlorothiophene

  • MDL number MFCD23135682
  • Cas Number : 83663-36-9
  • Molecular Weight : 231.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/83663-36-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/83663-36-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart