AMBH2D6F7E5A Ambeed, Inc.

Methyl 5-bromo-6-oxo-1,6-dihydropyridine-2-carboxylate

  • MDL number MFCD16036911
  • Cas Number : 178876-86-3
  • Molecular Weight : 232.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/178876-86-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/178876-86-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart