AMBH2D6F7E65 Ambeed, Inc.

tert-Butyl 3-hydroxy-3-methylpyrrolidine-1-carboxylate

  • MDL number MFCD13191771
  • Cas Number : 412278-02-5
  • Molecular Weight : 201.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/412278-02-5.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/412278-02-5.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart