AMBH2D6F7E99 Ambeed, Inc.

3-Fluoro-5-methylisonicotinic acid

  • MDL number MFCD18257382
  • Cas Number : 1211578-10-7
  • Molecular Weight : 155.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1211578-10-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1211578-10-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart