AMBH2D6F7EFB Ambeed, Inc.

(1H-Pyrrolo[2,3-b]pyridin-3-yl)methanol

  • MDL number MFCD11054203
  • Cas Number : 1065100-83-5
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1065100-83-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1065100-83-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart