AMBH2D6F7F19 Ambeed, Inc.

1-Methylcyclopropyl (4-nitrophenyl) carbonate

  • MDL number MFCD17014665
  • Cas Number : 1046817-22-4
  • Molecular Weight : 237.2100

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1046817-22-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1046817-22-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart