AMBH2D6F7F47 Ambeed, Inc.

Ethyl 4,5,6,7-tetrahydro-1H-indazole-5-carboxylate

  • MDL number MFCD21364708
  • Cas Number : 792848-34-1
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/792848-34-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/792848-34-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart