AMBH2D6F8001 Ambeed, Inc.

Benzo[d]thiazol-2-ylmethanamine

  • MDL number MFCD06660504
  • Cas Number : 42182-65-0
  • Molecular Weight : 164.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-127°C
Purity 95
Flash Point 124.4
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Shipping Temperature Normal
Country of Origin CN
Boiling point 282°C at 760 mmHg
Product Link https://www.ambeed.com/products/42182-65-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/42182-65-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart