AMBH2D6F80D8 Ambeed, Inc.

6-Chloro-4-iodo-3-methoxypyridazine

  • MDL number MFCD12827448
  • Cas Number : 181355-92-0
  • Molecular Weight : 270.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/181355-92-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/181355-92-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart