AMBH2D6F820C Ambeed, Inc.

2-Heptylquinolin-4(1H)-one

  • MDL number MFCD16619170
  • Cas Number : 40522-46-1
  • Molecular Weight : 243.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/40522-46-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/40522-46-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart