AMBH2D6F82C2 Ambeed, Inc.

Choline Fenofibrate

  • MDL number MFCD18382233
  • Cas Number : 856676-23-8
  • Molecular Weight : 421.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/856676-23-8.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/856676-23-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart