AMBH2D6F8386 Ambeed, Inc.

(2S,4R)-1-(tert-Butoxycarbonyl)-4-(2-iodobenzyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD06659436
  • Cas Number : 959573-29-6
  • Molecular Weight : 431.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/959573-29-6.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/959573-29-6.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart