AMBH2D6F8397 Ambeed, Inc.

PF-8380

  • MDL number MFCD20527274
  • Cas Number : 1144035-53-9
  • Molecular Weight : 478.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1144035-53-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1144035-53-9.html
pictograms GHS07
hstatements H302-H315-H319-H335-H413
Availability details for
Quantity
  • Availability
  • Add to Cart