AMBH2D6F8482 Ambeed, Inc.

(E)-N-(3,4-Dihydroxyphenethyl)-3-(4-hydroxyphenyl)acrylamide

  • MDL number MFCD17129089
  • Cas Number : 103188-46-1
  • Molecular Weight : 299.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/103188-46-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/103188-46-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart