AMBH2D6F8493 Ambeed, Inc.

H-Aib-OMe.HCl

  • MDL number MFCD00214247
  • Cas Number : 15028-41-8
  • Molecular Weight : 153.6100

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185
Synonym Methyl 2-amino-2-methylpropanoate hydrochloride
Purity 97
Flash Point 56.1
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/15028-41-8.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/15028-41-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart