AMBH2D6F84EB Ambeed, Inc.

2,6-Dibromo-4,8-bis((2-ethylhexyl)oxy)benzo[1,2-b:4,5-b']dithiophene

  • MDL number MFCD17018552
  • Cas Number : 1226782-13-3
  • Molecular Weight : 604.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 331.2
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 624.0�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1226782-13-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1226782-13-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart