AMBH2D6F8530 Ambeed, Inc.

5-Chloro-2,3-diphenylpyrazine

  • MDL number MFCD00234892
  • Cas Number : 41270-66-0
  • Molecular Weight : 266.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/41270-66-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/41270-66-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart