AMBH2D6F85D3 Ambeed, Inc.

4-Phenyl-3,4-dihydro-2H-benzo[e][1,3]oxazin-2-one

  • MDL number MFCD26406244
  • Cas Number : 1129278-73-4
  • Molecular Weight : 225.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 156.7
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 335.5�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1129278-73-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1129278-73-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart