AMBH2D6F85D6 Ambeed, Inc.

4-oxo-3,4-Dihydroquinazoline-6-carbonitrile

  • MDL number MFCD20039986
  • Cas Number : 117297-41-3
  • Molecular Weight : 171.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Hydroxyquinazoline-6-carbonitrile
Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/117297-41-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/117297-41-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart