AMBH2D6F85F3 Ambeed, Inc.

Ethyl 3-bromopropiolate

  • MDL number MFCD11977143
  • Cas Number : 41658-03-1
  • Molecular Weight : 177.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/41658-03-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/41658-03-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart