AMBH2D6F8638 Ambeed, Inc.

(Cyclohexylmethyl)boronic acid

  • MDL number MFCD09878538
  • Cas Number : 27762-64-7
  • Molecular Weight : 142.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-104°C
Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin China
Product Link https://www.ambeed.com/products/27762-64-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/27762-64-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart