AMBH2D6F866F Ambeed, Inc.

2-Chloro-5-methoxyquinoline

  • MDL number MFCD22394142
  • Cas Number : 160893-07-2
  • Molecular Weight : 193.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 141.6
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 310.5�0�t 760 mmHg
Product Link https://www.ambeed.com/products/160893-07-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/160893-07-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart