AMBH2D6F8674 Ambeed, Inc.

4,5-Dibromo-2H-1,2,3-triazole

  • MDL number MFCD19440717
  • Cas Number : 22300-52-3
  • Molecular Weight : 226.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/22300-52-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/22300-52-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart