AMBH2D6F86AC Ambeed, Inc.

6,6'-Dichloro-2,2'-bipyridine

  • MDL number MFCD11042483
  • Cas Number : 53344-72-2
  • Molecular Weight : 225.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220
Purity 97
Flash Point 199.445
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 354.681�t 760 mmHg
Product Link https://www.ambeed.com/products/53344-72-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/53344-72-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart