AMBH2D6F86D8 Ambeed, Inc.

Pyrimidine-4,6-dicarbonitrile

  • MDL number MFCD11870748
  • Cas Number : 50844-89-8
  • Molecular Weight : 130.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-95
Purity 95
Flash Point 108.2
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 293.8�0�t 760 mmHg
Product Link https://www.ambeed.com/products/50844-89-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/50844-89-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart