AMBH2D6F875A Ambeed, Inc.

Methyl 2,4,6-trichloronicotinate

  • MDL number MFCD21604740
  • Cas Number : 1218994-35-4
  • Molecular Weight : 240.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1218994-35-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1218994-35-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart