AMBH2D6F87D4 Ambeed, Inc.

3-Bromo-4-cyclopropylbenzoic acid

  • MDL number MFCD11110859
  • Cas Number : 1131622-50-8
  • Molecular Weight : 241.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1131622-50-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1131622-50-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart