AMBH2D6F87FA Ambeed, Inc.

3-Methoxypyridine-4-boronic acid

  • MDL number MFCD06801698
  • Cas Number : 1008506-24-8
  • Molecular Weight : 152.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1008506-24-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1008506-24-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart