AMBH2D6F881B Ambeed, Inc.

3-Bromo-4-fluoropyridin-2-amine

  • MDL number MFCD26743631
  • Cas Number : 1417407-29-4
  • Molecular Weight : 191.0011

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1417407-29-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/1417407-29-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart