AMBH2D6F8822 Ambeed, Inc.

6-Bromo-3-(trifluoromethyl)-1H-indazole

  • Cas Number : 1374258-63-5
  • Molecular Weight : 265.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1374258-63-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1374258-63-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart