AMBH2D6F8832 Ambeed, Inc.

5H-Benzo[d]benzo[4,5]imidazo[1,2-a]imidazole

  • MDL number MFCD24551866
  • Cas Number : 28890-99-5
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 193.9
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 397.0�0�t 760 mmHg
Product Link https://www.ambeed.com/products/28890-99-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/28890-99-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart