AMBH2D6F8856 Ambeed, Inc.

AZD7545

  • MDL number MFCD25976792
  • Cas Number : 252017-04-2
  • Molecular Weight : 478.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (R)-4-((3-Chloro-4-(3,3,3-trifluoro-2-hydroxy-2-methylpropanamido)phenyl)sulfonyl)-N,N-dimethylbenzamide
Purity 98
Flash Point 366.9
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 683.1�0�t 760 mmHg
Product Link https://www.ambeed.com/products/252017-04-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/252017-04-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart