AMBH2D6F8865 Ambeed, Inc.

3-(Naphthalen-1-yl)thiomorpholine

  • MDL number MFCD06797798
  • Cas Number : 914349-66-9
  • Molecular Weight : 229.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/914349-66-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/914349-66-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart