AMBH2D6F8896 Ambeed, Inc.

Methyl 4-chloro-3-(cyanomethyl)benzoate

  • MDL number MFCD09263465
  • Cas Number : 872091-83-3
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/872091-83-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/872091-83-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart