AMBH2D6F88DA Ambeed, Inc.

(4-Hydroxy-3-methoxy-5-nitrophenyl)(p-tolyl)methanone

  • MDL number MFCD12545750
  • Cas Number : 134612-80-9
  • Molecular Weight : 287.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/134612-80-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/134612-80-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart