AMBH2D6F88E3 Ambeed, Inc.

5,6-Dichloro-1H-pyrrolo[3,2-b]pyridin-2(3H)-one

  • MDL number MFCD11847072
  • Cas Number : 136888-26-1
  • Molecular Weight : 203.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/136888-26-1.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/136888-26-1.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart