AMBH2D6F8AA7 Ambeed, Inc.

4-Bromo-2,6-diiodophenol

  • MDL number MFCD00672916
  • Cas Number : 15459-51-5
  • Molecular Weight : 424.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/15459-51-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/15459-51-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart