AMBH2D6F8ABB Ambeed, Inc.

1-(2,6-Difluorobenzyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD09955379
  • Cas Number : 166196-11-8
  • Molecular Weight : 239.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/166196-11-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/166196-11-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart