AMBH2D6F8AC3 Ambeed, Inc.

(2R,3R,4S)-2-(Hydroxymethyl)tetrahydrofuran-3,4-diol

  • MDL number MFCD27923011
  • Cas Number : 491-19-0
  • Molecular Weight : 134.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/491-19-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/491-19-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart