AMBH2D6F8B0A Ambeed, Inc.

Ethyl 7-chloro-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate

  • MDL number MFCD19443838
  • Cas Number : 223141-46-6
  • Molecular Weight : 239.6600

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form Solid or liquid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/223141-46-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/223141-46-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart