AMBH2D6F8B14 Ambeed, Inc.

(2-Bromo-6-methoxypyridin-3-yl)methanol

  • MDL number N/A
  • Cas Number : 1807211-71-7
  • Molecular Weight : 218.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1807211-71-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1807211-71-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart