AMBH2D6F8B33 Ambeed, Inc.

5-Bromo-3-(methylthio)-1H-indazole

  • MDL number MFCD13183072
  • Cas Number : 201227-23-8
  • Molecular Weight : 243.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/201227-23-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/201227-23-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart