AMBH2D6F8B3E Ambeed, Inc.

(R)-2-Methoxypropanoic acid

  • MDL number MFCD01632582
  • Cas Number : 23943-96-6
  • Molecular Weight : 104.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/23943-96-6.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/23943-96-6.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart