AMBH2D6F8B72 Ambeed, Inc.

Benzyl 3-aminopiperidine-1-carboxylate hydrochloride

  • MDL number MFCD04038445
  • Cas Number : 960541-42-8
  • Molecular Weight : 270.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/960541-42-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/960541-42-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart