AMBH2D6F8BA4 Ambeed, Inc.

4-Chloro-1H-indazol-6-ol

  • MDL number MFCD07781949
  • Cas Number : 887569-87-1
  • Molecular Weight : 168.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/887569-87-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/887569-87-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart